The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174916.2 | NP_777576.1 | 1 - 2 | 151 | 95 |
GenBank mRNA | AK056441.1 | - | 1 - 2 | 101 | 95 |
AK292516.1 | - | 1 - 2 | 98 | 95 | |
AK302489.1 | - | 1 - 2 | 178 | 95 | |
AY061886.1 | - | 1 - 2 | 85 | 95 | |
BC113505.1 | - | 1 - 2 | 163 | 95 | |
BC113507.1 | - | 1 - 2 | 163 | 95 |