The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024847.2 | NP_001020018.1 | 3 - 4 | 711 | 98 |
NM_003242.5 | NP_003233.4 | 2 - 3 | 636 | 98 | |
XM_011534043.2 | XP_011532345.1 | 3 - 4 | 484 | 98 | |
XM_011534045.2 | XP_011532347.1 | 2 - 3 | 264 | 98 | |
XM_017007106.1 | XP_016862595.1 | 2 - 3 | 1850 | 98 | |
GenBank mRNA | AK300383.1 | - | 3 - 4 | 612 | 98 |
AK304404.1 | - | 3 - 4 | 380 | 98 | |
AK314102.1 | - | 2 - 3 | 552 | 98 | |
BC040499.1 | - | 2 - 3 | 637 | 98 | |
CB988859.1 | - | 3 - 4 | 639 | 98 | |
D28131.1 | - | 3 - 4 | 269 | 98 | |
KC792576.1 | - | 2 - 3 | 293 | 98 | |
KC792577.1 | - | 2 - 3 | 293 | 98 | |
KC792578.1 | - | 2 - 3 | 293 | 98 | |
KC792579.1 | - | 2 - 3 | 293 | 98 | |
KC792580.1 | - | 2 - 3 | 293 | 98 | |
KU178359.1 | - | 2 - 3 | 254 | 98 | |
KU178360.1 | - | 3 - 4 | 329 | 98 | |
M85079.1 | - | 2 - 3 | 589 | 98 |