The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015456.4 | NP_056271.3 | 6 - 7 | 1107 | 95 |
GenBank mRNA | AF464935.1 | - | 6 - 7 | 912 | 95 |
AK022651.1 | - | 4 - 5 | 603 | 95 | |
AK091056.1 | - | 6 - 7 | 1100 | 95 | |
AL050280.1 | - | 3 - 4 | 354 | 95 | |
BC011892.1 | - | 5 - 6 | 816 | 95 | |
BC019029.1 | - | 6 - 7 | 1066 | 95 |