The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145294.4 | NP_660337.3 | 15 - 16 | 1772 | 105 |
XM_017023023.1 | XP_016878512.1 | 15 - 16 | 1775 | 105 | |
GenBank mRNA | AB067511.1 | - | 13 - 14 | 3389 | 105 |
AK126622.1 | - | 1 - 2 | 411 | 105 | |
BC121185.1 | - | 15 - 16 | 1785 | 105 | |
BC121186.1 | - | 15 - 16 | 1782 | 105 |