The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145294.4 | NP_660337.3 | 9 - 10 | 1106 | 107 |
XM_017023023.1 | XP_016878512.1 | 9 - 10 | 1109 | 107 | |
GenBank mRNA | AB067511.1 | - | 8 - 9 | 1497 | 107 |
AK091062.1 | - | 9 - 10 | 1106 | 107 | |
BC121185.1 | - | 9 - 10 | 1119 | 107 | |
BC121186.1 | - | 9 - 10 | 1116 | 107 |