The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002363.4 | NP_002354.2 | 1 - 2 | 72 | 107 |
NM_177415.2 | NP_803134.1 | 1 - 2 | 72 | 107 | |
GenBank mRNA | AK314977.1 | - | 1 - 2 | 72 | 107 |
DB022312.1 | - | 1 - 2 | 72 | 107 |