Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF85
Entrez Gene ID
7639
Gene Name
zinc finger protein 85
Gene Aliases
HPF4, HTF1
Chromosome Location
Chr.19: 20923227 - 20950697 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256173.1 NP_001243102.1 2 - 3 245 111
NM_003429.4 NP_003420.2 3 - 4 372 111
GenBank mRNA BP199573.1 - 3 - 4 372 111
DB119881.1 - 2 - 3 194 111
DN994243.1 - 3 - 4 359 111
M61866.1 - 3 - 4 336 111
U35376.1 - 3 - 4 342 111

View Full Product Details