The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303051.1 | NP_001289980.1 | 3 - 4 | 560 | 78 |
NM_001303109.1 | NP_001290038.1 | 3 - 4 | 580 | 78 | |
NM_021633.3 | NP_067646.1 | 3 - 4 | 580 | 78 | |
XM_011509835.1 | XP_011508137.1 | 4 - 5 | 600 | 78 | |
XM_011509836.1 | XP_011508138.1 | 4 - 5 | 600 | 78 | |
XM_011509837.1 | XP_011508139.1 | 4 - 5 | 600 | 78 | |
XM_017001995.1 | XP_016857484.1 | 3 - 4 | 549 | 78 | |
GenBank mRNA | AF190900.1 | - | 3 - 4 | 504 | 78 |
AF306689.1 | - | 3 - 4 | 549 | 78 | |
AK027656.1 | - | 3 - 4 | 551 | 78 | |
AK223072.1 | - | 3 - 4 | 516 | 78 | |
AK301791.1 | - | 3 - 4 | 515 | 78 | |
BC003183.1 | - | 3 - 4 | 532 | 78 | |
BC004175.2 | - | 3 - 4 | 532 | 78 |