Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL12
Entrez Gene ID
59349
Gene Name
kelch like family member 12
Gene Aliases
C3IP1, DKIR
Chromosome Location
Chr.1: 202891096 - 202928644 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303051.1 NP_001289980.1 8 - 9 1339 127
NM_021633.3 NP_067646.1 8 - 9 1359 127
XM_011509835.1 XP_011508137.1 9 - 10 1379 127
XM_011509836.1 XP_011508138.1 9 - 10 1379 127
XM_017001995.1 XP_016857484.1 8 - 9 1328 127
GenBank mRNA AF190900.1 - 8 - 9 1283 127
AF306687.1 - 5 - 6 633 127
AK024412.1 - 3 - 4 450 127
AK027656.1 - 8 - 9 1330 127
AK223072.1 - 8 - 9 1295 127
AK301791.1 - 8 - 9 1294 127
BC003183.1 - 8 - 9 1311 127
BC004175.2 - 8 - 9 1311 127

View Full Product Details