Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXLNGY
Entrez Gene ID
246126
Gene Name
taxilin gamma pseudogene, Y-linked
Gene Aliases
CYorf15A, CYorf15B, TXLNG2P
Chromosome Location
Chr.Y: 19567358 - 19607170 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_045128.1 8 - 9 1172 155
GenBank mRNA AF332225.1 - 2 - 3 448 155
AL832583.2 - 3 - 4 4718 155
CD242693.1 - 3 - 4 515 155

View Full Product Details