Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTDNEP1
Entrez Gene ID
23399
Gene Name
CTD nuclear envelope phosphatase 1
Gene Aliases
DULLARD, HSA011916, NET56
Chromosome Location
Chr.17: 7243587 - 7251940 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143775.1 NP_001137247.1 1 - 2 484 104
NM_015343.4 NP_056158.2 2 - 3 551 104
GenBank mRNA AJ011916.1 - 1 - 2 134 104
AY364239.1 - 1 - 2 104 104
BC009295.2 - 2 - 3 550 104

View Full Product Details