The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009571.3 | NP_001009571.2 | 25 - 26 | 3805 | 102 |
NM_001167940.1 | NP_001161412.1 | 27 - 28 | 3940 | 102 | |
NM_017954.10 | NP_060424.9 | 27 - 28 | 3928 | 102 | |
XM_005250695.4 | XP_005250752.1 | 29 - 30 | 3693 | 102 | |
XM_005250696.4 | XP_005250753.1 | 28 - 29 | 3684 | 102 | |
XM_005250697.4 | XP_005250754.1 | 28 - 29 | 3684 | 102 | |
XM_005250698.4 | XP_005250755.1 | 28 - 29 | 3650 | 102 | |
XM_005250699.4 | XP_005250756.1 | 28 - 29 | 3672 | 102 | |
XM_005250700.4 | XP_005250757.1 | 27 - 28 | 3669 | 102 | |
XM_005250701.4 | XP_005250758.1 | 27 - 28 | 3663 | 102 | |
XM_005250702.4 | XP_005250759.1 | 26 - 27 | 3648 | 102 | |
XM_005250703.4 | XP_005250760.1 | 28 - 29 | 3573 | 102 | |
XM_005250704.4 | XP_005250761.1 | 27 - 28 | 3564 | 102 | |
XM_005250705.4 | XP_005250762.1 | 26 - 27 | 3549 | 102 | |
XM_005250706.4 | XP_005250763.1 | 26 - 27 | 3549 | 102 | |
XM_005250707.4 | XP_005250764.1 | 26 - 27 | 3543 | 102 | |
XM_005250708.3 | XP_005250765.1 | 24 - 25 | 3519 | 102 | |
XM_011516695.2 | XP_011514997.1 | 27 - 28 | 3530 | 102 | |
XM_011516696.2 | XP_011514998.1 | 26 - 27 | 3537 | 102 | |
XM_011516697.2 | XP_011514999.1 | 25 - 26 | 3528 | 102 | |
XM_017012794.1 | XP_016868283.1 | 26 - 27 | 3654 | 102 | |
XM_017012795.1 | XP_016868284.1 | 25 - 26 | 3540 | 102 | |
XM_017012796.1 | XP_016868285.1 | 30 - 31 | 3308 | 102 | |
XM_017012797.1 | XP_016868286.1 | 29 - 30 | 3359 | 102 | |
XM_017012798.1 | XP_016868287.1 | 28 - 29 | 3823 | 102 | |
XM_017012799.1 | XP_016868288.1 | 28 - 29 | 3610 | 102 | |
XM_017012800.1 | XP_016868289.1 | 26 - 27 | 2701 | 102 | |
GenBank mRNA | AB046811.1 | - | 23 - 24 | 2674 | 102 |
AF401638.1 | - | 25 - 26 | 3474 | 102 | |
AK000768.1 | - | 10 - 11 | 983 | 102 | |
AK025672.1 | - | 20 - 21 | 2313 | 102 | |
AK054853.1 | - | 12 - 13 | 1087 | 102 | |
AK126518.1 | - | 26 - 27 | 3407 | 102 | |
AL833058.1 | - | 22 - 23 | 2600 | 102 | |
AY264289.1 | - | 27 - 28 | 3532 | 102 | |
BC136601.1 | - | 25 - 26 | 3504 | 102 | |
BC144278.1 | - | 25 - 26 | 3504 | 102 | |
BC144279.1 | - | 27 - 28 | 3639 | 102 | |
KJ534798.1 | - | 26 - 27 | 3386 | 102 | |
KJ534800.1 | - | 24 - 25 | 3368 | 102 | |
KJ534801.1 | - | 25 - 26 | 3383 | 102 | |
KJ534802.1 | - | 25 - 26 | 3377 | 102 | |
KJ534803.1 | - | 26 - 27 | 3398 | 102 | |
KJ534804.1 | - | 27 - 28 | 3407 | 102 | |
KJ534805.1 | - | 24 - 25 | 3368 | 102 | |
KJ535060.1 | - | 13 - 14 | 1612 | 102 |