The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178510.1 | NP_848605.1 | 7 - 8 | 1088 | 106 |
XM_017017475.1 | XP_016872964.1 | 8 - 9 | 2895 | 106 | |
GenBank mRNA | AF487542.1 | - | 7 - 8 | 994 | 106 |
AJ541797.2 | - | 7 - 8 | 1088 | 106 |