The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270476.1 | NP_001257405.1 | 9 - 10 | 1221 | 90 |
NM_006245.3 | NP_006236.1 | 9 - 10 | 1204 | 90 | |
NM_180976.2 | NP_851307.1 | 9 - 10 | 1108 | 90 | |
NM_180977.2 | NP_851308.1 | 7 - 8 | 886 | 90 | |
GenBank mRNA | AB000634.1 | - | 9 - 10 | 1039 | 90 |
AB000635.1 | - | 7 - 8 | 733 | 90 | |
AB451342.1 | - | 9 - 10 | 1025 | 90 | |
AB451357.1 | - | 9 - 10 | 1025 | 90 | |
AK290604.1 | - | 9 - 10 | 1017 | 90 | |
AK291797.1 | - | 9 - 10 | 1112 | 90 | |
AK299696.1 | - | 9 - 10 | 1077 | 90 | |
BC001095.2 | - | 9 - 10 | 1088 | 90 | |
BC001175.1 | - | 9 - 10 | 1015 | 90 | |
BC010692.1 | - | 9 - 10 | 1098 | 90 | |
D78360.1 | - | 9 - 10 | 1006 | 90 | |
L76702.1 | - | 9 - 10 | 1212 | 90 |