The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282422.2 | NP_001269351.1 | 1 - 2 | 131 | 115 |
NM_005000.4 | NP_004991.1 | 1 - 2 | 131 | 115 | |
NR_104168.2 | 1 - 2 | 131 | 115 | ||
NR_104169.2 | 1 - 2 | 131 | 115 | ||
NR_111925.1 | 1 - 2 | 131 | 115 |