The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004687.4 | NP_004678.3 | 3 - 4 | 200 | 91 |
XM_005257784.2 | XP_005257841.1 | 2 - 3 | 349 | 91 | |
XM_005257785.4 | XP_005257842.1 | 4 - 5 | 317 | 91 | |
XM_005257786.4 | XP_005257843.1 | 3 - 4 | 1104 | 91 | |
XM_006722168.3 | XP_006722231.1 | 3 - 4 | 144 | 91 | |
XM_011525460.2 | XP_011523762.1 | 4 - 5 | 1197 | 91 | |
GenBank mRNA | AB014547.2 | - | 1 - 2 | 90 | 91 |
AF264717.1 | - | 3 - 4 | 143 | 91 | |
AK308877.1 | - | 2 - 3 | 349 | 91 | |
BC035609.1 | - | 3 - 4 | 161 | 91 | |
DR003190.1 | - | 3 - 4 | 221 | 91 |