The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004687.4 | NP_004678.3 | 9 - 10 | 968 | 73 |
XM_005257784.2 | XP_005257841.1 | 8 - 9 | 1117 | 73 | |
XM_005257785.4 | XP_005257842.1 | 10 - 11 | 1085 | 73 | |
XM_005257786.4 | XP_005257843.1 | 9 - 10 | 1872 | 73 | |
XM_006722168.3 | XP_006722231.1 | 9 - 10 | 912 | 73 | |
XM_011525460.2 | XP_011523762.1 | 10 - 11 | 1965 | 73 | |
GenBank mRNA | AB014547.2 | - | 7 - 8 | 858 | 73 |
AF264717.1 | - | 9 - 10 | 911 | 73 | |
BC035609.1 | - | 9 - 10 | 929 | 73 |