The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005474.4 | NP_005465.2 | 2 - 3 | 354 | 73 |
XM_005256906.4 | XP_005256963.1 | 2 - 3 | 313 | 73 | |
XM_017023990.1 | XP_016879479.1 | 2 - 3 | 343 | 73 | |
XM_017023995.1 | XP_016879484.1 | 2 - 3 | 349 | 73 | |
XM_017023996.1 | XP_016879485.1 | 2 - 3 | 348 | 73 | |
GenBank mRNA | AB011172.2 | - | 2 - 3 | 326 | 73 |
AK294760.1 | - | 2 - 3 | 322 | 73 | |
BC051824.1 | - | 2 - 3 | 233 | 73 | |
BM564861.1 | - | 2 - 3 | 351 | 73 |