Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPP4
Entrez Gene ID
58538
Gene Name
membrane palmitoylated protein 4
Gene Aliases
ALS2CR5, DLG6
Chromosome Location
Chr.2: 201644874 - 201700263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033066.2 NP_149055.1 8 - 9 870 70
XM_017004620.1 XP_016860109.1 8 - 9 2439 70
GenBank mRNA AB053302.1 - 8 - 9 870 70
AB053303.1 - 7 - 8 675 70
AF316032.1 - 8 - 9 870 70
AK131208.1 - 7 - 8 756 70
AK298589.1 - 7 - 8 818 70
AK298947.1 - 8 - 9 827 70
AK300622.1 - 7 - 8 756 70
BC132785.1 - 7 - 8 767 70
BC144206.1 - 7 - 8 767 70
BX647162.1 - 8 - 9 762 70
BX648531.1 - 7 - 8 644 70

View Full Product Details