Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPP4
Entrez Gene ID
58538
Gene Name
membrane palmitoylated protein 4
Gene Aliases
ALS2CR5, DLG6
Chromosome Location
Chr.2: 201644874 - 201700263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033066.2 NP_149055.1 9 - 10 941 107
XM_017004620.1 XP_016860109.1 9 - 10 2510 107
GenBank mRNA AB053303.1 - 8 - 9 746 107
AF316032.1 - 9 - 10 941 107
AK131208.1 - 8 - 9 827 107
AK298589.1 - 8 - 9 889 107
AK298947.1 - 9 - 10 898 107
AK300622.1 - 8 - 9 827 107
BC132785.1 - 8 - 9 838 107
BC144206.1 - 8 - 9 838 107
BX647162.1 - 9 - 10 833 107
BX648531.1 - 8 - 9 715 107

View Full Product Details