Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYT6
Entrez Gene ID
148281
Gene Name
synaptotagmin 6
Gene Aliases
sytVI
Chromosome Location
Chr.1: 114089292 - 114153919 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270805.1 NP_001257734.1 6 - 7 1515 70
NM_205848.3 NP_995320.1 6 - 7 1497 70
GenBank mRNA AK056448.1 - 6 - 7 1497 70
BC044948.1 - 6 - 7 1515 70

View Full Product Details