Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYRFL
Entrez Gene ID
196446
Gene Name
myelin regulatory factor-like
Gene Aliases
C12orf15, C12orf28, bcm1377
Chromosome Location
Chr.12: 69824518 - 69958726 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182530.2 NP_872336.2 14 - 15 1959 85
XM_011539052.2 XP_011537354.1 13 - 14 2440 85
XM_011539055.1 XP_011537357.1 12 - 13 1427 85
XM_017018967.1 XP_016874456.1 13 - 14 2241 85
XM_017018968.1 XP_016874457.1 14 - 15 2695 85
XM_017018969.1 XP_016874458.1 13 - 14 2175 85
XM_017018970.1 XP_016874459.1 12 - 13 1985 85
XM_017018971.1 XP_016874460.1 14 - 15 2764 85
XM_017018972.1 XP_016874461.1 14 - 15 2763 85
XM_017018973.1 XP_016874462.1 10 - 11 1656 85
GenBank mRNA AK057785.1 - 1 - 2 76 85

View Full Product Details