The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000191.2 | NP_000182.2 | 6 - 7 | 611 | 86 |
GenBank mRNA | AK122801.1 | - | 7 - 8 | 1164 | 86 |
AK313869.1 | - | 6 - 7 | 580 | 86 | |
BC010570.1 | - | 6 - 7 | 577 | 86 | |
BT009792.1 | - | 6 - 7 | 567 | 86 | |
CR456884.1 | - | 6 - 7 | 567 | 86 | |
L07033.1 | - | 6 - 7 | 581 | 86 |