Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRIN1
Entrez Gene ID
2902
Gene Name
glutamate ionotropic receptor NMDA type subunit 1
Gene Aliases
GluN1, MRD8, NMD-R1, NMDA1, NMDAR1, NR1
Chromosome Location
Chr.9: 137139092 - 137168762 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000832.6 NP_000823.4 17 - 18 2771 76
NM_001185090.1 NP_001172019.1 18 - 19 2834 76
NM_001185091.1 NP_001172020.1 18 - 19 2834 76
NM_007327.3 NP_015566.1 17 - 18 2771 76
NM_021569.3 NP_067544.1 17 - 18 2771 76
XM_005266071.3 XP_005266128.1 17 - 18 2441 76
XM_005266072.3 XP_005266129.1 18 - 19 2838 76
XM_005266073.4 XP_005266130.1 18 - 19 2899 76
XM_011518583.2 XP_011516885.1 18 - 19 2838 76
GenBank mRNA AB208918.1 - 17 - 18 2518 76
AB208999.1 - 18 - 19 2799 76
AF015730.1 - 18 - 19 2577 76
AF015731.1 - 18 - 19 2575 76
D13515.1 - 17 - 18 2612 76
L05666.1 - 17 - 18 2458 76
L13266.1 - 17 - 18 3534 76
L13267.1 - 12 - 13 1545 76
L13268.1 - 17 - 18 2411 76
U08106.1 - 11 - 12 1448 76

View Full Product Details