The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106.3 | NP_001097.2 | 7 - 8 | 983 | 101 |
XM_005265583.3 | XP_005265640.1 | 7 - 8 | 3445 | 101 | |
XM_017007514.1 | XP_016863003.1 | 7 - 8 | 3424 | 101 | |
XM_017007515.1 | XP_016863004.1 | 7 - 8 | 1069 | 101 | |
XM_017007516.1 | XP_016863005.1 | 7 - 8 | 4393 | 101 | |
GenBank mRNA | AB209449.1 | - | 6 - 7 | 4748 | 101 |
BC096243.1 | - | 7 - 8 | 963 | 101 | |
BC096244.1 | - | 7 - 8 | 963 | 101 | |
BC096245.1 | - | 6 - 7 | 1045 | 101 | |
BC099642.4 | - | 7 - 8 | 963 | 101 | |
X77533.1 | - | 7 - 8 | 963 | 101 |