The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002547.2 | NP_002538.1 | 10 - 11 | 1225 | 81 |
XM_005262270.1 | XP_005262327.1 | 10 - 11 | 1295 | 81 | |
XM_006724653.2 | XP_006724716.1 | 10 - 11 | 1295 | 81 | |
XM_011530961.1 | XP_011529263.1 | 10 - 11 | 1146 | 81 | |
XM_017029555.1 | XP_016885044.1 | 10 - 11 | 1295 | 81 | |
GenBank mRNA | AB102656.1 | - | 9 - 10 | 931 | 81 |
AJ001189.1 | - | 10 - 11 | 1569 | 81 | |
AK295769.1 | - | 10 - 11 | 1295 | 81 | |
BC140763.1 | - | 10 - 11 | 1263 | 81 |