Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FSTL3
Entrez Gene ID
10272
Gene Name
follistatin like 3
Gene Aliases
FLRG, FSRP
Chromosome Location
Chr.19: 676389 - 683392 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005860.2 NP_005851.1 1 - 2 137 114
GenBank mRNA AK291958.1 - 1 - 2 134 114
AY358917.1 - 1 - 2 118 114
BC005839.2 - 1 - 2 123 114
BC033119.1 - 1 - 2 109 114
BX439786.2 - 1 - 2 150 114
U76702.1 - 1 - 2 109 114

View Full Product Details