Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS3
Entrez Gene ID
9508
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 3
Gene Aliases
ADAMTS-4
Chromosome Location
Chr.4: 72280969 - 72568799 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014243.2 NP_055058.2 11 - 12 1640 124
XM_011532421.2 XP_011530723.1 11 - 12 1672 124
XM_011532422.2 XP_011530724.1 11 - 12 2171 124
GenBank mRNA AB002364.1 - 11 - 12 1592 124
BC130287.1 - 11 - 12 1637 124
BC132735.1 - 11 - 12 1637 124

View Full Product Details