Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDON
Entrez Gene ID
50937
Gene Name
cell adhesion associated, oncogene regulated
Gene Aliases
CDO, CDON1, HPE11, ORCAM
Chromosome Location
Chr.11: 125956813 - 126063335 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243597.1 NP_001230526.1 12 - 13 2494 86
NM_016952.4 NP_058648.4 12 - 13 2494 86
XM_011542862.2 XP_011541164.1 12 - 13 2655 86
XM_011542863.2 XP_011541165.1 12 - 13 2659 86
XM_011542864.2 XP_011541166.1 12 - 13 2572 86
XM_011542865.2 XP_011541167.1 12 - 13 2455 86
XM_011542866.2 XP_011541168.1 12 - 13 2655 86
XM_017017873.1 XP_016873362.1 12 - 13 2491 86
GenBank mRNA AB209396.1 - 3 - 4 883 86
AF004841.2 - 12 - 13 2522 86
BC098583.1 - 12 - 13 2494 86
BC114435.1 - 11 - 12 2398 86

View Full Product Details