The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144578.3 | NP_653179.1 | 3 - 4 | 892 | 88 |
XM_011537362.2 | XP_011535664.2 | 3 - 4 | 1736 | 88 | |
GenBank mRNA | AK025580.1 | - | 3 - 4 | 856 | 88 |
AL834285.1 | - | 3 - 4 | 875 | 88 | |
BC015621.2 | - | 3 - 4 | 853 | 88 |