Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44792348-44792593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLC
Entrez Gene ID
23624
Gene Name
Cbl proto-oncogene C
Gene Aliases
CBL-3, CBL-SL, RNF57
UniGene
Hs.466907
Gene Chromosome Location
Chr.19: 44777855-44800646 on Build GRCh38
Transcripts
NM_001130852.1 NM_012116.3 XM_005258696.3 XM_011526688.2 XM_011526689.2 XM_011526690.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
231958 c.1078G>A p.D360N Chr.19: 44792455-44792455 on Build GRCh38
6151515 c.1134G>T p.W378C Chr.19: 44792511-44792511 on Build GRCh38
319248 c.1022A>G p.Y341C Chr.19: 44792399-44792399 on Build GRCh38
5762846 c.1030A>G p.M344V Chr.19: 44792407-44792407 on Build GRCh38
5762845 c.1030A>G p.M344V Chr.19: 44792407-44792407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774164707 Chr.19: 44792421-44792421 on Build GRCh38
rs548133945 Chr.19: 44792417-44792417 on Build GRCh38
rs979268798 Chr.19: 44792460-44792460 on Build GRCh38
rs775236973 Chr.19: 44792428-44792428 on Build GRCh38
rs758907585 Chr.19: 44792389-44792389 on Build GRCh38
rs760268560 Chr.19: 44792431-44792431 on Build GRCh38
rs780773035 Chr.19: 44792549-44792550 on Build GRCh38
rs897758747 Chr.19: 44792383-44792383 on Build GRCh38
rs767651678 Chr.19: 44792552-44792552 on Build GRCh38
rs148655912 Chr.19: 44792447-44792447 on Build GRCh38
rs374379148 Chr.19: 44792474-44792474 on Build GRCh38
rs752110502 Chr.19: 44792540-44792540 on Build GRCh38
rs755448447 Chr.19: 44792551-44792551 on Build GRCh38
rs766921120 Chr.19: 44792465-44792465 on Build GRCh38
rs758885229 Chr.19: 44792455-44792455 on Build GRCh38
rs746891746 Chr.19: 44792502-44792502 on Build GRCh38
rs531086131 Chr.19: 44792472-44792472 on Build GRCh38
rs747076535 Chr.19: 44792405-44792405 on Build GRCh38
rs188263434 Chr.19: 44792526-44792526 on Build GRCh38
rs73570198 Chr.19: 44792564-44792564 on Build GRCh38
rs145265588 Chr.19: 44792442-44792442 on Build GRCh38
rs199898564 Chr.19: 44792535-44792535 on Build GRCh38
rs766605869 Chr.19: 44792538-44792538 on Build GRCh38
rs772681113 Chr.19: 44792530-44792530 on Build GRCh38
rs748321868 Chr.19: 44792416-44792416 on Build GRCh38
rs757061451 Chr.19: 44792556-44792557 on Build GRCh38
rs34873320 Chr.19: 44792475-44792475 on Build GRCh38
rs1005749994 Chr.19: 44792377-44792377 on Build GRCh38
rs60229589 Chr.19: 44792481-44792481 on Build GRCh38
rs759463367 Chr.19: 44792426-44792426 on Build GRCh38
rs371314325 Chr.19: 44792515-44792515 on Build GRCh38
rs1023454829 Chr.19: 44792448-44792448 on Build GRCh38
rs923737137 Chr.19: 44792470-44792470 on Build GRCh38
rs763654909 Chr.19: 44792443-44792443 on Build GRCh38
rs778634807 Chr.19: 44792478-44792478 on Build GRCh38
rs761439548 Chr.19: 44792520-44792520 on Build GRCh38
rs961495774 Chr.19: 44792399-44792399 on Build GRCh38
rs771700766 Chr.19: 44792484-44792484 on Build GRCh38
rs781426720 Chr.19: 44792412-44792412 on Build GRCh38
rs768327321 Chr.19: 44792506-44792506 on Build GRCh38
rs775597740 Chr.19: 44792499-44792499 on Build GRCh38
rs376189819 Chr.19: 44792555-44792555 on Build GRCh38
rs971614881 Chr.19: 44792471-44792471 on Build GRCh38
rs756093858 Chr.19: 44792556-44792556 on Build GRCh38
rs142187731 Chr.19: 44792438-44792438 on Build GRCh38
rs952137350 Chr.19: 44792441-44792441 on Build GRCh38
rs761530274 Chr.19: 44792446-44792446 on Build GRCh38
rs779284374 Chr.19: 44792373-44792373 on Build GRCh38
rs377703099 Chr.19: 44792476-44792476 on Build GRCh38
rs149074838 Chr.19: 44792407-44792407 on Build GRCh38
rs369690964 Chr.19: 44792391-44792391 on Build GRCh38
rs772036785 Chr.19: 44792427-44792427 on Build GRCh38
rs376434724 Chr.19: 44792381-44792381 on Build GRCh38
rs750794901 Chr.19: 44792451-44792451 on Build GRCh38

View Full Product Details