Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 34370754-34370979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNE2
Entrez Gene ID
9992
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 2
Gene Aliases
ATFB4, LQT5, LQT6, MIRP1
UniGene
Hs.551521
Gene Chromosome Location
Chr.21: 34364024-34371141 on Build GRCh38
Transcripts
NM_172201.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3550483 c.280G>A p.E94K Chr.21: 34370758-34370758 on Build GRCh38
1030438 c.305A>T p.N102I Chr.21: 34370783-34370783 on Build GRCh38
6889212 c.331C>A p.H111N Chr.21: 34370809-34370809 on Build GRCh38
4542189 c.313G>A p.E105K Chr.21: 34370791-34370791 on Build GRCh38
1413933 c.347C>T p.A116V Chr.21: 34370825-34370825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766924678 Chr.21: 34370879-34370879 on Build GRCh38
rs746625998 Chr.21: 34370845-34370845 on Build GRCh38
rs551483595 Chr.21: 34370911-34370911 on Build GRCh38
rs199473367 Chr.21: 34370825-34370825 on Build GRCh38
rs745612699 Chr.21: 34370830-34370830 on Build GRCh38
rs187917779 Chr.21: 34370832-34370832 on Build GRCh38
rs751463321 Chr.21: 34370909-34370909 on Build GRCh38
rs867563620 Chr.21: 34370834-34370834 on Build GRCh38
rs375557746 Chr.21: 34370896-34370896 on Build GRCh38
rs786205807 Chr.21: 34370840-34370840 on Build GRCh38
rs183427173 Chr.21: 34370795-34370795 on Build GRCh38
rs1050957877 Chr.21: 34370946-34370946 on Build GRCh38
rs962445137 Chr.21: 34370918-34370918 on Build GRCh38
rs16991657 Chr.21: 34370826-34370826 on Build GRCh38
rs776266940 Chr.21: 34370863-34370863 on Build GRCh38
rs751891377 Chr.21: 34370786-34370786 on Build GRCh38
rs201388192 Chr.21: 34370886-34370886 on Build GRCh38
rs1052615224 Chr.21: 34370891-34370891 on Build GRCh38
rs763545106 Chr.21: 34370781-34370781 on Build GRCh38
rs761420303 Chr.21: 34370865-34370865 on Build GRCh38
rs72550217 Chr.21: 34370882-34370882 on Build GRCh38
rs376886316 Chr.21: 34370828-34370828 on Build GRCh38
rs756561888 Chr.21: 34370842-34370843 on Build GRCh38
rs781438592 Chr.21: 34370796-34370796 on Build GRCh38
rs786205808 Chr.21: 34370849-34370849 on Build GRCh38
rs113659353 Chr.21: 34370789-34370789 on Build GRCh38
rs558860396 Chr.21: 34370861-34370861 on Build GRCh38
rs753438833 Chr.21: 34370779-34370779 on Build GRCh38
rs750011503 Chr.21: 34370830-34370830 on Build GRCh38
rs756380802 Chr.21: 34370817-34370817 on Build GRCh38
rs193181183 Chr.21: 34370883-34370883 on Build GRCh38
rs139202426 Chr.21: 34370835-34370835 on Build GRCh38
rs72550218 Chr.21: 34370912-34370912 on Build GRCh38
rs374093618 Chr.21: 34370867-34370867 on Build GRCh38
rs150790888 Chr.21: 34370824-34370824 on Build GRCh38
rs45610936 Chr.21: 34370847-34370848 on Build GRCh38
rs752888066 Chr.21: 34370810-34370810 on Build GRCh38
rs767047359 Chr.21: 34370782-34370782 on Build GRCh38
rs749615979 Chr.21: 34370872-34370873 on Build GRCh38

View Full Product Details