Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GATA4
Entrez Gene ID
2626
Gene Name
GATA binding protein 4
Gene Aliases
ASD2, TACHD, TOF, VSD1
Chromosome Location
Chr.8: 11676919 - 11760002 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308093.1 NP_001295022.1 2 - 3 1247 83
NM_001308094.1 NP_001295023.1 3 - 4 587 83
NM_002052.4 NP_002043.2 3 - 4 1398 83
XM_005272385.4 XP_005272442.1 4 - 5 1470 83
XM_005272386.1 XP_005272443.1 4 - 5 1658 83
XM_006716248.1 XP_006716311.1 4 - 5 1579 83
XM_011543817.2 XP_011542119.1 4 - 5 1649 83
XM_011543818.2 XP_011542120.1 3 - 4 1787 83
XM_017013312.1 XP_016868801.1 3 - 4 2103 83
GenBank mRNA AK097060.1 - 3 - 4 547 83
AY740706.1 - 2 - 3 790 83
BC101580.1 - 2 - 3 867 83
BC105108.1 - 2 - 3 865 83
BC143434.1 - 2 - 3 867 83
BC143479.1 - 2 - 3 870 83
D78260.1 - 2 - 3 1079 83
DA900813.1 - 2 - 3 328 83
L34357.1 - 2 - 3 1027 83

View Full Product Details