The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173806.3 | NP_776167.2 | 2 - 3 | 290 | 142 |
NR_033694.1 | 4 - 5 | 541 | 142 | ||
XM_006721029.3 | XP_006721092.1 | 3 - 4 | 494 | 142 | |
XM_011545785.2 | XP_011544087.1 | 3 - 4 | 497 | 142 | |
XM_017023109.1 | XP_016878598.1 | 2 - 3 | 316 | 142 | |
XM_017023110.1 | XP_016878599.1 | 4 - 5 | 555 | 142 | |
GenBank mRNA | BC021724.1 | - | 4 - 5 | 546 | 142 |
BC028605.1 | - | 4 - 5 | 546 | 142 | |
BC039562.1 | - | 4 - 5 | 538 | 142 | |
DB450393.1 | - | 2 - 3 | 277 | 142 |