The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304263.1 | NP_001291192.1 | 3 - 4 | 431 | 72 |
NM_174902.3 | NP_777562.1 | 4 - 5 | 578 | 72 | |
GenBank mRNA | AK075546.1 | - | 4 - 5 | 497 | 72 |
AK293930.1 | - | 3 - 4 | 373 | 72 | |
BC042754.1 | - | 4 - 5 | 481 | 72 | |
BC136286.1 | - | 4 - 5 | 578 | 72 | |
BC143824.1 | - | 3 - 4 | 431 | 72 |