Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CERS3
Entrez Gene ID
204219
Gene Name
ceramide synthase 3
Gene Aliases
ARCI9, LASS3
Chromosome Location
Chr.15: 100400395 - 100544982 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290341.2 NP_001277270.1 9 - 10 1213 151
NM_001290342.2 NP_001277271.1 8 - 9 1039 151
NM_001290343.1 NP_001277272.1 8 - 9 950 151
NM_178842.4 NP_849164.2 8 - 9 946 151
XM_011521355.2 XP_011519657.1 9 - 10 1256 151
XM_011521357.2 XP_011519659.1 9 - 10 1150 151
XM_011521358.1 XP_011519660.1 9 - 10 1034 151
XM_017022002.1 XP_016877491.1 8 - 9 1030 151
XM_017022003.1 XP_016877492.1 8 - 9 955 151
XM_017022004.1 XP_016877493.1 7 - 8 866 151
GenBank mRNA BC028703.1 - 8 - 9 151
BC034500.1 - 8 - 9 902 151
BC034970.1 - 8 - 9 948 151

View Full Product Details