The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181536.1 | NP_853514.1 | 8 - 9 | 1273 | 71 |
XM_017023201.1 | XP_016878690.1 | 8 - 9 | 1696 | 71 | |
XM_017023202.1 | XP_016878691.1 | 8 - 9 | 1696 | 71 | |
XM_017023203.1 | XP_016878692.1 | 8 - 9 | 1696 | 71 | |
XM_017023204.1 | XP_016878693.1 | 8 - 9 | 1696 | 71 | |
GenBank mRNA | AY164485.1 | - | 8 - 9 | 1273 | 71 |