The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181707.2 | NP_859058.2 | 5 - 6 | 712 | 142 |
XM_005257033.4 | XP_005257090.1 | 5 - 6 | 1052 | 142 | |
XM_005257034.4 | XP_005257091.1 | 5 - 6 | 1008 | 142 | |
XM_005257035.4 | XP_005257092.1 | 5 - 6 | 686 | 142 | |
XM_005257036.2 | XP_005257093.1 | 5 - 6 | 751 | 142 | |
XM_017024166.1 | XP_016879655.1 | 5 - 6 | 698 | 142 | |
GenBank mRNA | BC036356.1 | - | 5 - 6 | 707 | 142 |
BC048806.1 | - | 5 - 6 | 704 | 142 | |
CD742737.1 | - | 1 - 2 | 142 |