Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABHD12B
Entrez Gene ID
145447
Gene Name
abhydrolase domain containing 12B
Gene Aliases
BEM46L3, C14orf29, c14_5314
Chromosome Location
Chr.14: 50872090 - 50904970 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206673.1 NP_001193602.1 4 - 5 471 85
NM_181533.3 NP_853511.2 3 - 4 343 85
NM_181814.1 NP_861535.1 2 - 3 240 85
XM_006720052.1 XP_006720115.1 2 - 3 237 85
XM_011536472.1 XP_011534774.1 4 - 5 541 85
XM_011536474.2 XP_011534776.1 4 - 5 992 85
XM_017021031.1 XP_016876520.1 3 - 4 405 85
GenBank mRNA AK128464.1 - 2 - 3 237 85
AY311396.1 - 4 - 5 471 85
AY311397.1 - 3 - 4 343 85
AY311398.1 - 2 - 3 240 85
BC034603.1 - 2 - 3 207 85
BC107141.1 - 2 - 3 240 85
BC107142.1 - 2 - 3 238 85

View Full Product Details