The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206673.1 | NP_001193602.1 | 8 - 9 | 718 | 66 |
NM_181533.3 | NP_853511.2 | 7 - 8 | 590 | 66 | |
NM_181814.1 | NP_861535.1 | 6 - 7 | 487 | 66 | |
XM_006720052.1 | XP_006720115.1 | 6 - 7 | 484 | 66 | |
XM_011536472.1 | XP_011534774.1 | 8 - 9 | 788 | 66 | |
XM_011536474.2 | XP_011534776.1 | 8 - 9 | 1239 | 66 | |
XM_017021031.1 | XP_016876520.1 | 7 - 8 | 652 | 66 | |
GenBank mRNA | AK128464.1 | - | 6 - 7 | 484 | 66 |
AY311396.1 | - | 8 - 9 | 718 | 66 | |
AY311397.1 | - | 7 - 8 | 590 | 66 | |
AY311398.1 | - | 6 - 7 | 487 | 66 | |
AY311399.1 | - | 5 - 6 | 414 | 66 | |
BC034603.1 | - | 6 - 7 | 454 | 66 | |
BC107141.1 | - | 6 - 7 | 487 | 66 | |
BC107142.1 | - | 6 - 7 | 485 | 66 |