The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182539.3 | NP_872345.2 | 3 - 4 | 1003 | 59 |
XM_005248872.1 | XP_005248929.1 | 3 - 4 | 1455 | 59 | |
XM_005248873.2 | XP_005248930.1 | 3 - 4 | 1455 | 59 | |
XM_005248874.2 | XP_005248931.1 | 3 - 4 | 714 | 59 | |
XM_006715009.3 | XP_006715072.1 | 4 - 5 | 1119 | 59 | |
XM_011514336.2 | XP_011512638.1 | 3 - 4 | 1276 | 59 | |
XM_011514338.2 | XP_011512640.1 | 4 - 5 | 1509 | 59 | |
XM_011514339.2 | XP_011512641.1 | 3 - 4 | 725 | 59 | |
GenBank mRNA | AK301825.1 | - | 3 - 4 | 1003 | 59 |
BC035022.1 | - | 3 - 4 | 975 | 59 |