The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178470.4 | NP_848565.2 | 2 - 2 | 2961 | 108 |
GenBank mRNA | BC035674.1 | - | 2 - 2 | 2953 | 108 |