Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLK9
Entrez Gene ID
284366
Gene Name
kallikrein related peptidase 9
Gene Aliases
KLK-L3, KLKL3
Chromosome Location
Chr.19: 51002513 - 51009634 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012315.1 NP_036447.1 3 - 4 560 100
GenBank mRNA AY551001.1 - 2 - 3 403 100

View Full Product Details