Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM15
Entrez Gene ID
89870
Gene Name
tripartite motif containing 15
Gene Aliases
RNF93, ZNF178, ZNFB7
Chromosome Location
Chr.6: 30163206 - 30172696 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033229.2 NP_150232.2 1 - 1 737 78
XM_011514988.2 XP_011513290.1 1 - 1 258 78
GenBank mRNA AF220132.1 - 1 - 1 258 78
AF220133.1 - 1 - 1 258 78
AK290280.1 - 1 - 1 443 78
AK291853.1 - 1 - 1 395 78
BC038585.1 - 1 - 1 338 78
U34249.1 - 1 - 1 751 78

View Full Product Details