Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44777855-44778102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLC
Entrez Gene ID
23624
Gene Name
Cbl proto-oncogene C
Gene Aliases
CBL-3, CBL-SL, RNF57
UniGene
Hs.466907
Gene Chromosome Location
Chr.19: 44777855-44800646 on Build GRCh38
Transcripts
NM_001130852.1 NM_012116.3 XM_005258696.3 XM_011526688.2 XM_011526689.2 XM_011526690.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3535746 c.166C>T p.R56* Chr.19: 44778097-44778097 on Build GRCh38
3103348 c.128C>T p.S43L Chr.19: 44778059-44778059 on Build GRCh38
7219223 c.115G>T p.V39L Chr.19: 44778046-44778046 on Build GRCh38
3422920 c.109C>T p.L37L Chr.19: 44778040-44778040 on Build GRCh38
3971083 c.133C>T p.R45W Chr.19: 44778064-44778064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751204879 Chr.19: 44777902-44777902 on Build GRCh38
rs558512943 Chr.19: 44778045-44778045 on Build GRCh38
rs866606097 Chr.19: 44778032-44778032 on Build GRCh38
rs778906584 Chr.19: 44778022-44778022 on Build GRCh38
rs896172136 Chr.19: 44777933-44777933 on Build GRCh38
rs759152258 Chr.19: 44777981-44777981 on Build GRCh38
rs528411573 Chr.19: 44777877-44777877 on Build GRCh38
rs770372319 Chr.19: 44777971-44777971 on Build GRCh38
rs762570407 Chr.19: 44777898-44777898 on Build GRCh38
rs567104270 Chr.19: 44777878-44777878 on Build GRCh38
rs776435797 Chr.19: 44777887-44777887 on Build GRCh38
rs761041888 Chr.19: 44778011-44778011 on Build GRCh38
rs772312798 Chr.19: 44777942-44777942 on Build GRCh38
rs761887572 Chr.19: 44777891-44777891 on Build GRCh38
rs765850610 Chr.19: 44777900-44777900 on Build GRCh38
rs983411997 Chr.19: 44778056-44778056 on Build GRCh38
rs767823003 Chr.19: 44777903-44777903 on Build GRCh38
rs931299718 Chr.19: 44778064-44778064 on Build GRCh38
rs544784225 Chr.19: 44778036-44778036 on Build GRCh38
rs756006315 Chr.19: 44778042-44778042 on Build GRCh38
rs764374254 Chr.19: 44778013-44778013 on Build GRCh38
rs776955522 Chr.19: 44778080-44778080 on Build GRCh38
rs750480822 Chr.19: 44778023-44778023 on Build GRCh38
rs1020514306 Chr.19: 44777997-44777997 on Build GRCh38
rs200348346 Chr.19: 44778008-44778008 on Build GRCh38
rs758611319 Chr.19: 44778025-44778025 on Build GRCh38
rs536079345 Chr.19: 44777896-44777896 on Build GRCh38
rs762946724 Chr.19: 44777968-44777968 on Build GRCh38
rs754372194 Chr.19: 44778018-44778018 on Build GRCh38
rs1000666675 Chr.19: 44777999-44777999 on Build GRCh38
rs201826947 Chr.19: 44777912-44777912 on Build GRCh38
rs773799426 Chr.19: 44778054-44778054 on Build GRCh38
rs749288188 Chr.19: 44778046-44778046 on Build GRCh38
rs756681255 Chr.19: 44777913-44777913 on Build GRCh38
rs975094314 Chr.19: 44778030-44778030 on Build GRCh38
rs747946824 Chr.19: 44777952-44777952 on Build GRCh38
rs1057366149 Chr.19: 44777904-44777904 on Build GRCh38
rs757709044 Chr.19: 44778019-44778019 on Build GRCh38
rs1032029318 Chr.19: 44777947-44777947 on Build GRCh38
rs937568625 Chr.19: 44777879-44777879 on Build GRCh38
rs754284335 Chr.19: 44777917-44777917 on Build GRCh38
rs773891788 Chr.19: 44777980-44777980 on Build GRCh38
rs902236013 Chr.19: 44777973-44777973 on Build GRCh38
rs775127808 Chr.19: 44778076-44778076 on Build GRCh38
rs767195128 Chr.19: 44778005-44778005 on Build GRCh38
rs140237650 Chr.19: 44777942-44777943 on Build GRCh38
rs769820375 Chr.19: 44777892-44777892 on Build GRCh38
rs771859187 Chr.19: 44778013-44778014 on Build GRCh38
rs780884827 Chr.19: 44777944-44777944 on Build GRCh38
rs778239531 Chr.19: 44777916-44777916 on Build GRCh38
rs778966197 Chr.19: 44777918-44777918 on Build GRCh38
rs762265193 Chr.19: 44778083-44778083 on Build GRCh38
rs886978806 Chr.19: 44777885-44777885 on Build GRCh38
rs147885389 Chr.19: 44778055-44778055 on Build GRCh38
rs34932144 Chr.19: 44777954-44777954 on Build GRCh38
rs148948759 Chr.19: 44778077-44778077 on Build GRCh38
rs779009321 Chr.19: 44777927-44777927 on Build GRCh38
rs746098134 Chr.19: 44777941-44777941 on Build GRCh38
rs757331996 Chr.19: 44777920-44777920 on Build GRCh38
rs773289730 Chr.19: 44777965-44777965 on Build GRCh38
rs780339606 Chr.19: 44778031-44778031 on Build GRCh38

View Full Product Details