Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1352977-1353250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UVSSA
Entrez Gene ID
57654
Gene Name
UV stimulated scaffold protein A
Gene Aliases
KIAA1530, UVSS3
UniGene
Hs.380475
Gene Chromosome Location
Chr.4: 1345662-1388049 on Build GRCh38
Transcripts
NM_001317934.1 NM_001317935.1 NM_020894.3 XM_017008490.1 XM_017008491.1 XM_017008492.1 XM_017008493.1 XM_017008494.1 XM_017008495.1 XM_017008496.1 XM_017008497.1 XM_017008498.1 XM_017008499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1186505 c.625G>T p.D209Y Chr.4: 1353104-1353104 on Build GRCh38
6166484 c.607C>T p.L203L Chr.4: 1353086-1353086 on Build GRCh38
1186506 c.703G>T p.V235L Chr.4: 1353182-1353182 on Build GRCh38
5090158 c.551-6C>T p.? Chr.4: 1353024-1353024 on Build GRCh38
7107757 c.637G>C p.E213Q Chr.4: 1353116-1353116 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749890443 Chr.4: 1353162-1353162 on Build GRCh38
rs769842022 Chr.4: 1353081-1353081 on Build GRCh38
rs140577099 Chr.4: 1353195-1353195 on Build GRCh38
rs776359468 Chr.4: 1353151-1353151 on Build GRCh38
rs144339897 Chr.4: 1353025-1353025 on Build GRCh38
rs917566736 Chr.4: 1353132-1353132 on Build GRCh38
rs778183745 Chr.4: 1353174-1353174 on Build GRCh38
rs760575642 Chr.4: 1353145-1353145 on Build GRCh38
rs765958000 Chr.4: 1353204-1353204 on Build GRCh38
rs778329600 Chr.4: 1353177-1353177 on Build GRCh38
rs148799777 Chr.4: 1353161-1353161 on Build GRCh38
rs771953897 Chr.4: 1353137-1353137 on Build GRCh38
rs144411768 Chr.4: 1353121-1353121 on Build GRCh38
rs77892061 Chr.4: 1353115-1353115 on Build GRCh38
rs996421348 Chr.4: 1353179-1353179 on Build GRCh38
rs763602199 Chr.4: 1353103-1353103 on Build GRCh38
rs753243616 Chr.4: 1353050-1353050 on Build GRCh38
rs150061207 Chr.4: 1352998-1352998 on Build GRCh38
rs775424662 Chr.4: 1353138-1353138 on Build GRCh38
rs745950800 Chr.4: 1353134-1353134 on Build GRCh38
rs925140740 Chr.4: 1353112-1353112 on Build GRCh38
rs560564580 Chr.4: 1353199-1353199 on Build GRCh38
rs758683465 Chr.4: 1353222-1353222 on Build GRCh38
rs535069303 Chr.4: 1353093-1353093 on Build GRCh38
rs776433683 Chr.4: 1353192-1353192 on Build GRCh38
rs141286641 Chr.4: 1353060-1353060 on Build GRCh38
rs763916276 Chr.4: 1353147-1353147 on Build GRCh38
rs755488475 Chr.4: 1353113-1353113 on Build GRCh38
rs143547791 Chr.4: 1353108-1353108 on Build GRCh38
rs148246275 Chr.4: 1353056-1353056 on Build GRCh38
rs200478324 Chr.4: 1353051-1353051 on Build GRCh38
rs745895956 Chr.4: 1353182-1353182 on Build GRCh38
rs760545821 Chr.4: 1353102-1353102 on Build GRCh38
rs367736394 Chr.4: 1353087-1353087 on Build GRCh38
rs777483270 Chr.4: 1353045-1353045 on Build GRCh38
rs954895838 Chr.4: 1353099-1353099 on Build GRCh38
rs753225382 Chr.4: 1353002-1353002 on Build GRCh38
rs137963993 Chr.4: 1353104-1353104 on Build GRCh38
rs146853550 Chr.4: 1353175-1353175 on Build GRCh38
rs750015341 Chr.4: 1353111-1353111 on Build GRCh38
rs749520493 Chr.4: 1353126-1353126 on Build GRCh38
rs150774883 Chr.4: 1353092-1353092 on Build GRCh38
rs764512705 Chr.4: 1353213-1353213 on Build GRCh38
rs778613098 Chr.4: 1353053-1353053 on Build GRCh38
rs773109686 Chr.4: 1353010-1353010 on Build GRCh38
rs762126831 Chr.4: 1353007-1353007 on Build GRCh38
rs750609200 Chr.4: 1353221-1353221 on Build GRCh38
rs374789666 Chr.4: 1353058-1353058 on Build GRCh38
rs780171750 Chr.4: 1353231-1353231 on Build GRCh38
rs376480468 Chr.4: 1353032-1353032 on Build GRCh38
rs371958204 Chr.4: 1353157-1353157 on Build GRCh38
rs771114453 Chr.4: 1353127-1353127 on Build GRCh38
rs772113626 Chr.4: 1353183-1353183 on Build GRCh38
rs763098474 Chr.4: 1353012-1353012 on Build GRCh38
rs147131553 Chr.4: 1353109-1353109 on Build GRCh38
rs548572482 Chr.4: 1353014-1353014 on Build GRCh38
rs773021607 Chr.4: 1353198-1353198 on Build GRCh38
rs779295093 Chr.4: 1353220-1353220 on Build GRCh38
rs751520782 Chr.4: 1353019-1353019 on Build GRCh38
rs577559437 Chr.4: 1353152-1353152 on Build GRCh38
rs537542108 Chr.4: 1353084-1353084 on Build GRCh38
rs1020776398 Chr.4: 1353107-1353107 on Build GRCh38
rs768088458 Chr.4: 1353167-1353167 on Build GRCh38
rs527603101 Chr.4: 1353039-1353039 on Build GRCh38
rs761196386 Chr.4: 1353210-1353210 on Build GRCh38
rs779133494 Chr.4: 1353178-1353178 on Build GRCh38
rs1010398560 Chr.4: 1353077-1353077 on Build GRCh38
rs375343086 Chr.4: 1353024-1353024 on Build GRCh38
rs775470552 Chr.4: 1353184-1353184 on Build GRCh38
rs781765925 Chr.4: 1353114-1353114 on Build GRCh38
rs375280783 Chr.4: 1353194-1353194 on Build GRCh38
rs748541217 Chr.4: 1353061-1353061 on Build GRCh38
rs886579502 Chr.4: 1353029-1353029 on Build GRCh38
rs768536941 Chr.4: 1353187-1353187 on Build GRCh38
rs904755870 Chr.4: 1353100-1353100 on Build GRCh38
rs549659618 Chr.4: 1353089-1353089 on Build GRCh38
rs761185498 Chr.4: 1353212-1353212 on Build GRCh38
rs751086316 Chr.4: 1353211-1353211 on Build GRCh38
rs150060662 Chr.4: 1353120-1353120 on Build GRCh38
rs76217876 Chr.4: 1353216-1353216 on Build GRCh38
rs762894740 Chr.4: 1353086-1353086 on Build GRCh38
rs756645260 Chr.4: 1353173-1353173 on Build GRCh38
rs144329202 Chr.4: 1353215-1353215 on Build GRCh38
rs562897525 Chr.4: 1352999-1352999 on Build GRCh38
rs115978884 Chr.4: 1353172-1353172 on Build GRCh38
rs774357492 Chr.4: 1353128-1353128 on Build GRCh38
rs201508186 Chr.4: 1353186-1353186 on Build GRCh38
rs758014008 Chr.4: 1353166-1353166 on Build GRCh38
rs17164682 Chr.4: 1353008-1353008 on Build GRCh38
rs776678516 Chr.4: 1353169-1353171 on Build GRCh38

View Full Product Details