Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SKA2
Entrez Gene ID
348235
Gene Name
spindle and kinetochore associated complex subunit 2
Gene Aliases
FAM33A
Chromosome Location
Chr.17: 59109947 - 59155439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182620.3 NP_872426.1 3 - 4 576 105
GenBank mRNA AK056473.1 - 3 - 4 328 105
BC017873.1 - 3 - 4 337 105
BC106003.1 - 3 - 4 301 105

View Full Product Details