The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145313.2 | NP_001138785.1 | 9 - 10 | 1087 | 89 |
NM_001287191.1 | NP_001274120.1 | 8 - 9 | 991 | 89 | |
NM_001287192.1 | NP_001274121.1 | 8 - 9 | 991 | 89 | |
XM_005252254.2 | XP_005252311.1 | 9 - 10 | 1320 | 89 | |
XM_005252257.3 | XP_005252314.1 | 8 - 9 | 986 | 89 | |
XM_005252260.3 | XP_005252317.1 | 7 - 8 | 953 | 89 | |
XM_011519077.2 | XP_011517379.1 | 10 - 11 | 978 | 89 | |
XM_011519078.2 | XP_011517380.2 | 9 - 10 | 985 | 89 | |
XM_011519079.2 | XP_011517381.2 | 9 - 10 | 987 | 89 | |
XM_011519080.2 | XP_011517382.1 | 3 - 4 | 1683 | 89 | |
XM_011519081.2 | XP_011517383.1 | 3 - 4 | 1668 | 89 | |
XM_017015182.1 | XP_016870671.1 | 9 - 10 | 1320 | 89 | |
XM_017015183.1 | XP_016870672.1 | 8 - 9 | 986 | 89 | |
XM_017015184.1 | XP_016870673.1 | 4 - 5 | 496 | 89 | |
XM_017015185.1 | XP_016870674.1 | 9 - 10 | 993 | 89 | |
XM_017015186.1 | XP_016870675.1 | 3 - 4 | 336 | 89 | |
XM_017015187.1 | XP_016870676.1 | 8 - 9 | 986 | 89 | |
GenBank mRNA | AK296433.1 | - | 8 - 9 | 902 | 89 |
AK299350.1 | - | 9 - 10 | 1019 | 89 | |
AK299491.1 | - | 8 - 9 | 902 | 89 | |
AK309817.1 | - | 9 - 10 | 1328 | 89 | |
BC036746.1 | - | 3 - 4 | 430 | 89 |