The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163264.1 | NP_001156736.1 | 3 - 4 | 515 | 102 |
NM_001163265.1 | NP_001156737.1 | 3 - 4 | 572 | 102 | |
NM_004845.4 | NP_004836.2 | 3 - 4 | 572 | 102 | |
XM_017029977.1 | XP_016885466.1 | 4 - 5 | 397 | 102 | |
GenBank mRNA | AF052510.1 | - | 3 - 4 | 399 | 102 |
AF148464.1 | - | 3 - 4 | 343 | 102 | |
AK226121.1 | - | 3 - 4 | 515 | 102 | |
AK296333.1 | - | 3 - 4 | 466 | 102 | |
AK315323.1 | - | 3 - 4 | 572 | 102 | |
BC045634.1 | - | 3 - 4 | 330 | 102 |