Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SPDYA
- Entrez Gene ID
-
245711
- Gene Name
- speedy/RINGO cell cycle regulator family member A
- Gene Aliases
-
RINGO3, RINGOA, SPDY1, SPY1
- Chromosome Location
-
Chr.2: 28810834 - 28850610 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_182756.3 |
NP_877433.2 |
2 - 3 |
190 |
66 |
View Full Product Details